HALO® ES-C18, BIOCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
93412-102 93413-102 93414-102 93410-402 93410-602 93410-702 93411-402 93411-602 93411-702 93412-202 93412-302 93412-402 93412-502 93412-602 93412-702 93412-902 93413-202 93413-302 93413-402 93413-502 93413-602 93413-702 93413-902 93414-202 93414-302 93414-402 93414-502 93414-602 93414-702 93414-902 94315-402 94315-602 94315-702 94316-402 94316-602 94316-702
76443-522PK 1124.96 CAD
76443-522 76443-528 76443-532 76445-092 76445-096 76445-100 76445-104 76445-108 76445-112 76445-116 76445-120 76445-124 76445-128 76445-132 76445-136 76445-142 76445-146 76445-152 76445-158 76445-162 76445-166 76445-170 76445-174 76445-178 76445-182 76445-186 76445-190 76445-194 76445-198 76445-204 76445-208 76445-214 76445-220 76445-224 76445-228 76445-232 76445-236 76445-240 76445-244 76445-248 76445-252 76445-256 76445-260 76445-264 76445-268 76445-272 76445-276 76445-280 76446-740
HALO® ES-C18, BIOCLASS, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® BIOCLASS ES-C18 400 Å Protein columns made with innovative large pore Fused-Core® particle technology provide narrower peaks and better recoveries for high-resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. HALO 400 Å columns are optimized for high performance in HPLC, UHPLC and LCMS large molecule applications, delivering excellent reproducibility and column lifetimes.
  • Ideal for large protein analysis such as mAbs and polypeptides
  • Well suited for high speed protein separations
  • Outstanding low bleed
More Product Information