2-8CELLsius Short-Term Storage Solutions, Protide Pharmaceuticals
Supplier: PROTIDE PHARMACEUTICALS
PP338-100 PP338-500 PP338-1000 PP338-1000B
76443-304EA 200.63 CAD
76443-304 76443-306 76443-308 76443-310
2-8CELLsius Short-Term Storage Solutions, Protide Pharmaceuticals
Cell Culture Supplements and Additives
Ready to use, balanced, cytoprotective, protein free solution with bio-stabilizers for short-term storage, regenerative medicine, stem cell processing and transportation of biological material. Short term storage can be achieved at 2-8°C or at ambient temperature for 24-48 hours. Does not contain any growth factors, hormones, or antibiotics. 2-8CELLsius™ can also be utilized as a cell washing solution.
  • TYPE II drug master file
  • USP grade components/Highest grade
  • Supports high cell viability
  • No animal or human proteins
  • No growth factors, hormones, antibiotics
  • Ready to use/sterile membrane filtered
Certifications: cGMP manufactured.