Intelsius® BioTherm (15...25 °C) Sample Transport Shippers, DGP Intelsius
Supplier: DGP INTELSIUS LLC
BT085 BT086 BT087 BT088 BT105
76441-448EA 136.65 CAD
76441-448 76441-450 76441-452 76441-454 76441-464
Intelsius® BioTherm (15...25 °C) Sample Transport Shippers, DGP Intelsius
Mailers and Shippers
Intelsius® BioTherm shippers are suitable for shipping temperature critical samples requiring a stable room temperature of between 15 and 25 °C.
  • Passive cooling
  • Compliant labels
  • Reusable cold pack technology
  • Regulatory compliant packaging
The BioTherm range of high performance 15 to 25 °C solutions are fully compliant to IATA, ADR, and DOT (49 CFR) transport regulations. These systems are qualified to ensure sample integrity for up to 96 hours against 15 to 25 °C temperature exposures. Each system is provided with handles and the correct complianceMore Product Information