QUANTIFOIL® R 1.2 Holey Carbon Films on Grids, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
Q2100AR1.3 Q2100AR1.3-2NM Q2100CR1.3-2NM Q210AR1.3 Q210AR1.3-2NM Q210AR21.2 Q210CR1.3 Q210CR1.3-2NM Q210CR21.2 Q210NR1.3 Q210NR21.2 Q225AR1.3 Q225AR1.3-2NM Q225AR21.2 Q225CR1.3-2NM Q225CR21.2 Q225NR21.2 Q250AR1.3-2NM Q250AR21.2 Q250CR1.3-2NM Q250CR21.2 Q250NR21.2 Q3100AR1.3 Q3100AR1.3-2NM Q3100CR1.3-2NM Q310AR1.3 Q310AR1.3-2NM Q310AR21.2 Q310CR1.3 Q310CR1.3-2NM Q310CR21.2 Q310NR1.3 Q310NR21.2 Q325AR1.3 Q325AR1.3-2NM Q325AR21.2 Q325CR1.3-2NM Q325CR21.2 Q325NR21.2 Q350AR1.3 Q350AR1.3-2NM Q350AR21.2 Q350CR1.3-2NM Q350CR21.2 Q350NR21.2 Q4100AR1.3 Q4100AR1.3-2NM Q4100CR1.3-2NM Q410AR1.3 Q410AR1.3-2NM Q410AR21.2 Q410CR1.3 Q410CR1.3-2NM Q410CR21.2 Q410NR1.3 Q410NR21.2 Q425AR1.3-2NM Q425AR21.2 Q425CR1.3-2NM Q425CR21.2 Q425NR21.2 Q450AR1.3-2NM Q450AR21.2 Q450CR1.3-2NM Q450CR21.2 Q450NR21.2
76440-108PK 4499.92 CAD
76440-108 76440-110 76440-122 76440-144 76440-146 76440-164 76440-198 76440-200 76440-218 76440-252 76440-268 76440-318 76440-320 76440-334 76440-358 76440-372 76440-402 76440-422 76440-436 76440-460 76440-474 76440-504 76440-524 76440-526 76440-538 76440-560 76440-562 76440-580 76440-614 76440-616 76440-634 76440-668 76440-684 76440-734 76440-736 76440-750 76440-770 76440-784 76440-814 76440-832 76440-834 76440-848 76440-868 76440-882 76440-912 76440-930 76440-932 76440-944 76440-966 76440-968 76440-986 76441-020 76441-022 76441-040 76441-074 76441-090 76441-140 76441-154 76441-174 76441-188 76441-218 76441-236 76441-250 76441-270 76441-284 76441-314
QUANTIFOIL® R 1.2 Holey Carbon Films on Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
QUANTIFOIL® is a carbon film for electron microscopy or low-energy electron point source microscopy.
  • Mounted on copper, nickel or gold grids
  • Foil is 12 nm thick
  • Thicknesses between 10 and 25 nm upon request
QUANTIFOIL® is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL® with circular holes is used in cryo-electron tomographic reconstruction. The roundness of the holes isMore Product Information