QUANTIFOIL® R 3.5 Holey Carbon Films on Grids
Supplier: ELECTRON MICROSCOPY SCIENCE
Q210AR-35 Q210CR-35 Q210NR-35 Q220AR-35 Q220CR-35 Q220NR-35 Q310AR-35 Q310CR-35 Q310NR-35 Q320AR-35 Q320CR-35 Q320NR-35 Q410AR-35 Q410CR-35 Q410NR-35 Q420AR-35 Q420CR-35 Q420NR-35
76440-170PK 610.37 CAD
76440-170 76440-224 76440-272 76440-296 76440-304 76440-312 76440-586 76440-640 76440-688 76440-712 76440-720 76440-728 76440-992 76441-046 76441-094 76441-118 76441-126 76441-134
QUANTIFOIL® R 3.5 Holey Carbon Films on Grids
Graticules, Reticles and Grids
QUANTIFOIL® is a carbon film for electron microscopy or low-energy electron point source microscopy.
  • Preferred over foils with smaller holes if the carbon film should be outside the frame of the image
  • Mounted on copper, nickel or gold grids
  • Foil is 12 nm thick
  • Thicknesses between 10 and 25 nm upon
QUANTIFOIL® is a perforated support foil with pre-defined hole size, shape and arrangement that has advantages in electron microscopy (EM) or low-energy electron point source (LEEPS) when compared with conventional holey film. QUANTIFOIL® with circular holesMore Product Information