Quantifoil® Holey Carbon Films, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Q225-AR2 Q225-CR2 Q225-AS7 Q225-CR1 Q225-NMA Q250-NR1 Q250-NR2 Q225-AR1 Q225-CR4 Q225-CS7 Q250-AR2 Q250-AS7 Q250-CR4 Q250-NMA Q225-CMA Q225-NR4 Q250-AR1 Q250-CR2 Q250-CS7 Q225-CR1.3 Q250-NR1.3 Q225CR-35 Q325CR1.3 Q350CR1.3 Q3100CR1.3 Q425CR1.3 Q450CR1.3 Q4100CR1.3 Q3100CMA Q325CR-35 Q250AR-14 Q325CR2 Q350CR2 Q3100CR2 Q350AR2 Q3100AR2 Q425CR2 Q450CR2 Q2100CR1 Q425CR1 Q450CR1 Q3100CR-4 Q325CR-06 Q450AR-06 Q4100CR-4 Q2100CR2 Q2100CR5 Q3100CR5 Q3100CR1010 Q4100CR1010 Q2100CR1020 Q3100CR1020 Q4100CR1020 Q2100CR175 Q3100CR175 Q4100CR175 Q3100CR-12 Q4100CR-12 Q2100CR21.2 Q3100CR21.2 Q4100CR21.2 Q2100CR3 Q3100CR3 Q4100CR3 Q2100CR510 Q3100CR510 Q4100CR510 Q4100AR105 Q2100AR1010 Q3100AR1010 Q2100NR-12 Q3100NR-12 Q4100NR-12 Q2100NR21.2 Q3100NR21.2 Q4100NR21.2 Q2100NR3 Q4100NR3 Q2100NR5 Q3100NR5 Q4100NR5 Q2100NR510 Q3100NR510 Q4100NR510 Q2100AR-12 Q3100AR-12 Q4100AR-12 Q2100AR3 Q3100AR3 Q4100AR3 Q2100AR5 Q3100AR5 Q4100AR5 Q2100AR510 Q3100AR510 Q3100CR665 Q4100CR665 Q2100CR6100 Q3100CR6100 Q4100CR6100 Q2100CR105 Q3100CR105 Q4100CR105 Q2100CR1010 Q2100NR665 Q3100NR665 Q4100NR665 Q3100NR6100 Q4100NR6100 Q2100NR105 Q3100NR105 Q4100NR105 Q2100NR1010 Q4100NR1010 Q2100NR1020 Q3100NR1020 Q4100NR1020 Q2100NR175 Q3100NR175 Q4100NR175 Q2100AR665 Q3100AR665 Q4100AR665 Q2100AR6100 Q3100AR6100 Q4100AR6100 Q2100AR105 Q4100AR1010 Q2100AR1020 Q3100AR1020 Q2100AR175 Q3100AR175 Q210AR-06 Q210CR-06 Q210NR-06 Q220AR-06 Q220CR-06 Q220NR-06 Q310AR-06 Q310CR-06 Q310NR-06 Q320AR-06 Q320CR-06 Q320NR-06 Q410AR-06 Q410CR-06 Q410NR-06 Q420AR-06 Q420CR-06 Q420NR-06
100491-204EA 1136.86 CAD
100491-204 100491-216 100491-206 100491-212 100491-222 100491-264 100491-268 100491-202 100491-218 100491-220 100491-240 100491-242 100491-254 100491-258 100491-208 100491-234 100491-238 100491-252 100491-256 102101-506 102101-512 102773-134 102773-148 102773-150 102773-152 102773-160 102773-162 102773-164 102773-222 102773-286 102773-320 102773-350 102773-352 102773-354 102773-364 102773-366 102773-368 102773-370 102773-386 102773-410 102773-412 102773-428 102773-456 102773-478 102773-488 102773-502 103305-800 103305-802 103305-820 103305-822 103305-824 103305-826 103305-828 103305-830 103305-832 103305-834 103306-344 103306-346 103306-354 103306-356 103306-404 103306-406 103306-408 103306-410 103306-418 103306-420 103306-422 103306-574 103306-576 103306-578 103306-810 103306-812 103306-814 103306-822 103306-824 103306-826 103306-828 103306-832 103306-834 103306-836 103306-838 103306-840 103306-842 103306-844 103306-892 103306-894 103306-896 103306-898 103306-900 103306-902 103306-904 103306-906 103306-908 103306-910 103306-912 103306-918 103306-920 103306-922 103306-924 103306-926 103306-928 103306-930 103306-932 103306-934 103306-992 103306-994 103306-996 103307-000 103307-002 103307-004 103307-006 103307-008 103307-010 103307-014 103307-016 103307-018 103307-020 103307-022 103307-024 103307-026 103307-030 103307-032 103307-034 103307-036 103307-038 103307-040 103307-042 103307-044 103307-046 103307-048 103307-052 103307-054 76440-140 76440-194 76440-248 76440-292 76440-300 76440-308 76440-556 76440-610 76440-664 76440-708 76440-716 76440-724 76440-962 76441-016 76441-070 76441-114 76441-122 76441-130
Quantifoil® Holey Carbon Films, Electron Microscopy Sciences
Graticules, Reticles and Grids
Holey carbon film for electron microscopy or low-energy electron point source microscopy.
  • Offered with circular and square, orthogonal arranged holes
  • Hole sizes from 0.6, 1, 1.2, 2, 3, 3.5, 5, 6, 10 and 17.5 µm
  • 200, 300, and 400 mesh copper and nickel grids
  • Carbon film is about 10 to 12 nm thick
The surface properties of QUANTIFOIL® holey carbon support film, especially the wetting properties, may have to be adapted according to one's particular requirements. Untreated aging QUANTIFOIL® tends to be hydrophobic. Hydrophillicity of the foil can beMore Product Information