Graphene Oxide TEM Support Films, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
GOHC300CU10 GOHC300CU25 GOHC300CU50 GOLC300CU10 GOLC300CU100 GOLC300CU25 GOLC300CU50 GOQ200R24CU10 GOQ200R24CU25 GOQ200R24CU50 GOQ300R22CU10 GOQ300R22CU25 GOQ300R24CU10 GOQ300R24CU25 GOQ300R24CU50 GOQ400R1213100 GOQ400R1213A10 GOQ400R1213A25 GOQ400R1213AU50 GOQ400R1213C10 GOQ400R1213C25 GOQ400R1213C50
76439-700PK 434.15 CAD
76439-700 76439-702 76439-704 76439-706 76439-708 76439-710 76439-712 76439-714 76439-716 76439-718 76439-720 76439-722 76439-724 76439-726 76439-728 76439-730 76439-732 76439-734 76439-736 76439-738 76439-740 76439-742
Graphene Oxide TEM Support Films, Electron Microscopy Sciences
Graticules, Reticles and Grids
Graphene Oxide (GO) support film is a super thin (<1 nm), naturally hydrophilic layer placed over the Holey, Lacey or Quantifoil support film on copper or gold grids.
  • Works well with Holey Carbon, Lacey Carbon and Quantifoil grid types, effectively spanning the gaps
  • Less expensive to produce due to complexity of graphene manufacturing
  • Better background contrast than graphene, results in higher resolution
Pre-treatment of GO Support Films is unnecessary. By default, the hydrophilic surface spreads particles evenly across the grid. A hydrophobicMore Product Information