Pure Silicon TEM Windows, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
76042-71 76042-72 76042-73 76042-74 76042-75 76042-76 76042-77
76439-590PK 1426.57 CAD
76439-590 76439-592 76439-594 76439-596 76439-598 76439-600 76439-602 76439-604 76439-606
Pure Silicon TEM Windows, Electron Microscopy Sciences
Graticules, Reticles and Grids
Pure silicon TEM windows feature imaging windows with 5 to 35 nm thickness, reducing background contribution and interference for higher contrast imaging. Most impressively, 5 nm thick non-porous pure silicon TEM Windows are thinner than the thinnest commercially available amorphous carbon membranes.
  • Plasma cleanable - can be vigorously plasma cleaned to remove organic contamination, unlike traditional carbon grids
  • Silicon composition - Sputter-deposited, pure, intrinsic silicon
  • Minimal background signal - Enables elemental analyses of samples containing
More Product Information