StepSaver™ Individual Extraction Stations, Environmental Express®
Supplier: Environmental Express
G6000 G6001 G6047 G6048 G6090 G6091
76436-978EA 3452.77 CAD
76436-978 76436-980 76436-982 76436-984 76436-986 76436-988
StepSaver™ Individual Extraction Stations, Environmental Express®
Extractors Automatic Extractor Systems
Constructed out of durable PVC and plastic materials, these compact, single-place extraction stations make oil and grease extractions easy.
  • Can be daisy-chained together to run multiple samples at one
  • Meets EPA method 1664 requirements for oil and grease extractions
  • Two separate chambers help keep water out of your extract
They are designed to be used for any type of Solid Phase Extraction (SPE) technique using an extraction disk. Single stations can be daisy-chained together to run multiple samples at once for faster sample throughput.
StepSaver™More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable