Sulfuric acid concentrate 0.2 N, Snip & Pour™

Supplier: ENVIRONMENTAL EXPRESS
Danger

C7010
76436-846EA 222.78 CAD
76436-846
Sulfuric acid concentrate 0.2 N, Snip & Pour™
Sulfuric acid

Deliver precise volume with pre-measured reagents.


  • Easy-to-use, just snip and pour into receiver tube
  • Increase accuracy by eliminating the need to measure and pipette
  • Packaged in convenient PFA tubes

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR