CE MicroIntensive™ G Duo Mop, Vileda Professional
Supplier: VWR International
76436-786
76436-786CS 1232.26 CAD
76436-786
CE MicroIntensive™ G Duo Mop, Vileda Professional
Mops
Single-use mop, superior results. Double-sided mop is twice as efficient as single-sided mops.
  • High performance polyester-microfiber for excellent cleaning result
  • High surface coverage with dual-sided usage
  • Independently proven bacteria reduction of 99.99%
  • Saves time, money, and chemical use
  • Excellent residue remover on glass and stainless steel
  • ""G"" version is pre-laundered, gamma irradiated, double-bagged, lot # coded with expiration date
Vileda Professional® is proud to announce a new microfiber mop for those looking to simplify and speed-up theMore Product Information