Avantor® Hichrom HI-WAX, GC Columns
Supplier: VWR International
76430-948 76430-950 76430-952 76430-954 76430-956 76430-958 76430-960 76430-962 76430-964 76430-966 76430-968 76430-970 76430-972 76430-974 76430-976 76430-978 76430-980 76431-424 76431-426 76431-428 76431-430 76435-750 76435-752 76435-754 76435-756 76435-758 76435-760 76435-762 76435-764 76435-766 76435-768 76435-770 76435-772 76435-774 76435-776 76435-778 76435-780 76435-782 76435-784 76435-786 76435-788 76435-790 76435-792 76435-794 76435-796 76435-798 76435-800 76435-802 76435-804 76435-806 76435-808 76435-810 76435-812 76435-814 76435-816 76435-818 76435-820 76435-822 76435-824 76435-826 76435-828 76435-830 76435-832 76435-834 76435-836 76435-838 76435-840 76435-842 76435-844 76435-846 76435-848 76435-850 76435-852 76435-854 76435-856 76435-858 76435-860 76435-862 76435-864 76435-866 76435-868 76435-870 76435-872 76435-874 76435-876 76435-878 76435-880 76435-882 76435-884 76435-886 76435-888 76435-890 76435-892 76435-894 76435-896 76435-898 76435-900 76435-902 76435-904 76435-906 76435-908 76435-910 76435-912 76435-914 76435-916 76435-918 76435-920 76435-922 76435-924 76435-926 76435-928 76435-930 76435-932 76435-934 76435-936 76435-938 76435-940 76435-942 76435-944 76435-946 76435-948 76435-950 76435-952 76435-954 76435-956 76435-958 76435-960 76435-962 76435-964 76435-966 76435-968 76435-970 76435-972 76435-974 76435-976 76435-978 76435-980 76435-982 76435-984 76435-986 76435-988 76435-990 76435-992 76435-994 76435-996 76435-998 76436-000 76436-002 76436-004 76436-006
76430-948EA 1266.04 CAD
76430-948 76430-950 76430-952 76430-954 76430-956 76430-958 76430-960 76430-962 76430-964 76430-966 76430-968 76430-970 76430-972 76430-974 76430-976 76430-978 76430-980 76431-424 76431-426 76431-428 76431-430 76435-750 76435-752 76435-754 76435-756 76435-758 76435-760 76435-762 76435-764 76435-766 76435-768 76435-770 76435-772 76435-774 76435-776 76435-778 76435-780 76435-782 76435-784 76435-786 76435-788 76435-790 76435-792 76435-794 76435-796 76435-798 76435-800 76435-802 76435-804 76435-806 76435-808 76435-810 76435-812 76435-814 76435-816 76435-818 76435-820 76435-822 76435-824 76435-826 76435-828 76435-830 76435-832 76435-834 76435-836 76435-838 76435-840 76435-842 76435-844 76435-846 76435-848 76435-850 76435-852 76435-854 76435-856 76435-858 76435-860 76435-862 76435-864 76435-866 76435-868 76435-870 76435-872 76435-874 76435-876 76435-878 76435-880 76435-882 76435-884 76435-886 76435-888 76435-890 76435-892 76435-894 76435-896 76435-898 76435-900 76435-902 76435-904 76435-906 76435-908 76435-910 76435-912 76435-914 76435-916 76435-918 76435-920 76435-922 76435-924 76435-926 76435-928 76435-930 76435-932 76435-934 76435-936 76435-938 76435-940 76435-942 76435-944 76435-946 76435-948 76435-950 76435-952 76435-954 76435-956 76435-958 76435-960 76435-962 76435-964 76435-966 76435-968 76435-970 76435-972 76435-974 76435-976 76435-978 76435-980 76435-982 76435-984 76435-986 76435-988 76435-990 76435-992 76435-994 76435-996 76435-998 76436-000 76436-002 76436-004 76436-006
Avantor® Hichrom HI-WAX, GC Columns
Chromatography Columns
Very popular PEG high polarity phase for a wide range of applications, including (for example): essential oils, flavors and fragrances, alkenols.
  • Phase composition: crossbonded polyethylene glycol (PEG)
  • Long lifetime
Max temperature limits may change depending on the film thickness.