Avantor® Hichrom HI-5, GC Columns
Supplier: VWR International
76430-410 76430-412 76430-414 76430-416 76430-418 76430-420 76430-422 76430-424 76430-426 76430-428 76430-430 76430-432 76430-434 76430-436 76430-438 76430-440 76430-442 76431-172 76431-174 76431-176 76431-178 76431-180 76431-182 76431-184 76434-912 76434-914 76434-916 76434-918 76434-920 76434-922 76434-924 76434-926 76434-928 76434-930 76434-932 76434-934 76434-936 76434-938 76434-940 76434-942 76434-944 76434-946 76434-948 76434-950 76434-952 76434-954 76434-956 76434-958 76434-960 76434-962 76434-964 76434-966 76434-968 76434-970 76434-972 76434-974 76434-976 76434-978 76434-980 76434-982 76434-984 76434-986 76434-988 76434-990 76434-992 76434-994 76434-996 76434-998 76435-000 76435-002 76435-004 76435-006 76435-008 76435-010 76435-012 76435-014 76435-016 76435-018 76435-020 76435-022 76435-024 76435-026 76435-028 76435-030 76435-032 76435-034 76435-036 76435-038 76435-040 76435-042 76435-044 76435-046 76435-048 76435-050 76435-052 76435-054 76435-056 76435-058 76435-060 76435-062 76435-064 76435-066 76435-068 76435-070 76435-072 76435-074 76435-076 76435-078 76435-080 76435-082 76435-084 76435-086 76435-088 76435-090 76435-092 76435-094 76435-096 76435-098 76435-100 76435-102 76435-104 76435-106 76435-108 76435-110 76435-112 76435-114 76435-116 76435-118 76435-120 76435-122 76435-124 76435-126 76435-128 76435-130 76435-132 76435-134 76435-136 76435-138 76435-140 76435-142 76435-144 76435-146 76435-148 76435-150 76435-152 76435-154 76435-156 76435-158 76435-160 76435-162 76435-164 76435-166 76435-168 76435-170 76435-172 76435-174 76435-176 76435-178 76435-180 76435-182 76435-184 76435-186 76435-188 76435-190 76435-192
76430-410EA 1292.42 CAD
76430-410 76430-412 76430-414 76430-416 76430-418 76430-420 76430-422 76430-424 76430-426 76430-428 76430-430 76430-432 76430-434 76430-436 76430-438 76430-440 76430-442 76431-172 76431-174 76431-176 76431-178 76431-180 76431-182 76431-184 76434-912 76434-914 76434-916 76434-918 76434-920 76434-922 76434-924 76434-926 76434-928 76434-930 76434-932 76434-934 76434-936 76434-938 76434-940 76434-942 76434-944 76434-946 76434-948 76434-950 76434-952 76434-954 76434-956 76434-958 76434-960 76434-962 76434-964 76434-966 76434-968 76434-970 76434-972 76434-974 76434-976 76434-978 76434-980 76434-982 76434-984 76434-986 76434-988 76434-990 76434-992 76434-994 76434-996 76434-998 76435-000 76435-002 76435-004 76435-006 76435-008 76435-010 76435-012 76435-014 76435-016 76435-018 76435-020 76435-022 76435-024 76435-026 76435-028 76435-030 76435-032 76435-034 76435-036 76435-038 76435-040 76435-042 76435-044 76435-046 76435-048 76435-050 76435-052 76435-054 76435-056 76435-058 76435-060 76435-062 76435-064 76435-066 76435-068 76435-070 76435-072 76435-074 76435-076 76435-078 76435-080 76435-082 76435-084 76435-086 76435-088 76435-090 76435-092 76435-094 76435-096 76435-098 76435-100 76435-102 76435-104 76435-106 76435-108 76435-110 76435-112 76435-114 76435-116 76435-118 76435-120 76435-122 76435-124 76435-126 76435-128 76435-130 76435-132 76435-134 76435-136 76435-138 76435-140 76435-142 76435-144 76435-146 76435-148 76435-150 76435-152 76435-154 76435-156 76435-158 76435-160 76435-162 76435-164 76435-166 76435-168 76435-170 76435-172 76435-174 76435-176 76435-178 76435-180 76435-182 76435-184 76435-186 76435-188 76435-190 76435-192
Avantor® Hichrom HI-5, GC Columns
Chromatography Columns
Versatile, general purpose, low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 is a crossbonded phase and assure a long lifetime even under high temperature limits.
Max temperature limits may change depending on the film thickness.