Avantor® Hichrom HI-1, GC Columns
Supplier: VWR International
76430-094 76430-096 76430-098 76430-100 76430-102 76430-104 76430-106 76430-108 76430-110 76430-112 76430-114 76430-116 76430-118 76430-120 76430-122 76430-124 76430-126 76431-050 76431-052 76431-054 76431-056 76434-154 76434-156 76434-158 76434-160 76434-162 76434-164 76434-166 76434-168 76434-170 76434-172 76434-174 76434-176 76434-178 76434-180 76434-182 76434-184 76434-186 76434-188 76434-190 76434-192 76434-194 76434-196 76434-198 76434-200 76434-202 76434-204 76434-206 76434-208 76434-210 76434-212 76434-214 76434-216 76434-218 76434-220 76434-222 76434-224 76434-226 76434-228 76434-230 76434-232 76434-234 76434-236 76434-238 76434-240 76434-242 76434-244 76434-246 76434-248 76434-250 76434-252 76434-254 76434-256 76434-258 76434-260 76434-262 76434-264 76434-266 76434-268 76434-270 76434-272 76434-274 76434-276 76434-278 76434-280 76434-282 76434-284 76434-286 76434-288 76434-290 76434-292 76434-294 76434-296 76434-298 76434-300 76434-302 76434-304 76434-306 76434-308 76434-310 76434-312 76434-314 76434-316 76434-318 76434-320 76434-322 76434-324 76434-326 76434-328 76434-330 76434-332 76434-334 76434-336 76434-338 76434-340 76434-342 76434-344 76434-346 76434-348 76434-350 76434-352 76434-354 76434-356 76434-358 76434-360 76434-362 76434-364 76434-366 76434-368 76434-370 76434-372 76434-374 76434-376 76434-378 76434-380 76434-382 76434-384 76434-386 76434-388 76434-390 76434-392 76434-394 76434-396 76434-398 76434-400 76434-402 76434-404 76434-406 76434-408 76434-410 76434-412 76434-414 76434-416 76434-418 76434-420 76434-422 76434-424 76434-426
76430-094EA 1266.04 CAD
76430-094 76430-096 76430-098 76430-100 76430-102 76430-104 76430-106 76430-108 76430-110 76430-112 76430-114 76430-116 76430-118 76430-120 76430-122 76430-124 76430-126 76431-050 76431-052 76431-054 76431-056 76434-154 76434-156 76434-158 76434-160 76434-162 76434-164 76434-166 76434-168 76434-170 76434-172 76434-174 76434-176 76434-178 76434-180 76434-182 76434-184 76434-186 76434-188 76434-190 76434-192 76434-194 76434-196 76434-198 76434-200 76434-202 76434-204 76434-206 76434-208 76434-210 76434-212 76434-214 76434-216 76434-218 76434-220 76434-222 76434-224 76434-226 76434-228 76434-230 76434-232 76434-234 76434-236 76434-238 76434-240 76434-242 76434-244 76434-246 76434-248 76434-250 76434-252 76434-254 76434-256 76434-258 76434-260 76434-262 76434-264 76434-266 76434-268 76434-270 76434-272 76434-274 76434-276 76434-278 76434-280 76434-282 76434-284 76434-286 76434-288 76434-290 76434-292 76434-294 76434-296 76434-298 76434-300 76434-302 76434-304 76434-306 76434-308 76434-310 76434-312 76434-314 76434-316 76434-318 76434-320 76434-322 76434-324 76434-326 76434-328 76434-330 76434-332 76434-334 76434-336 76434-338 76434-340 76434-342 76434-344 76434-346 76434-348 76434-350 76434-352 76434-354 76434-356 76434-358 76434-360 76434-362 76434-364 76434-366 76434-368 76434-370 76434-372 76434-374 76434-376 76434-378 76434-380 76434-382 76434-384 76434-386 76434-388 76434-390 76434-392 76434-394 76434-396 76434-398 76434-400 76434-402 76434-404 76434-406 76434-408 76434-410 76434-412 76434-414 76434-416 76434-418 76434-420 76434-422 76434-424 76434-426
Avantor® Hichrom HI-1, GC Columns
Chromatography Columns
Really versatile general purpose apolar phase for a wide range of applications, including (for example): PCBs, simulated distillation, gases, essential oils, hydrocarbons analysis, pesticides, semivolatiles, oxygenates. HI-1 is a crossbonded phase and assure a long lifetime even under high temperature limits.
  • Phase composition: Crossbonded 100% Methyl Polysiloxane (100% Dimethylpolysiloxane)
  • Long lifetime
Max temperature limits may change depending on the film thickness.