Avantor® Hichrom HI-WAX MS, GC Columns
Supplier: VWR International
76431-440
76431-442
76431-444
76431-446
76433-594
76433-596
76433-598
76433-600
76433-602
76433-604
76433-606
76433-608
76433-610
76433-612
76433-614
76433-616
76433-618
76433-620
76433-622
76433-624
76433-626
76433-628
76433-630
76433-632
76433-634
76433-636
76433-638
76433-640
76433-642
76433-644
76433-646
76433-648
76433-650
76433-652
76433-654
76433-656
76433-658
76433-660
76433-662
76433-664
76433-666
76433-668
76433-670
76433-672
76433-674
76433-676
76433-678
76433-680
76433-682
76433-684
76433-686
76433-688
76433-690
76433-692
76433-694
76433-696
76433-698
76433-700
76433-702
76433-704
76433-706
76433-708
76433-710
76433-712
76433-714
76433-716
76433-718
76433-720
76433-722
76433-724
76433-726
76433-728
76433-730
76433-732
76433-734
76433-736
76433-738
76433-740
76433-742
76433-744
76433-746
76433-748
76433-750
76433-752
76433-754
76433-756
76433-758
76433-760
76433-762
76433-764
76433-766
76433-768
76433-770
76433-772
76433-774
76433-776
76433-778
76433-780
76433-782
76433-784
76433-786
76433-788
76433-790
76433-792
76433-794
76433-796
76433-798
76433-800
76433-802
76433-804
76433-806
76433-808
76433-810
76433-812
76433-814
76433-816
76433-818
76433-820
76433-822
76433-824
76433-826
76433-828
76433-830
76433-832
76433-834
76433-836
76433-838
76433-840
76433-842
76433-844
76433-846
76433-848
76433-850
76434-120
76434-122
76434-124
76434-126
76434-128
76434-130
76434-132
76434-134
76434-136
76434-138
76434-140
76434-142
76434-144
76434-146
76434-148
76434-150
76434-152
76431-440EA
1637.34
CAD
76431-440
76431-442
76431-444
76431-446
76433-594
76433-596
76433-598
76433-600
76433-602
76433-604
76433-606
76433-608
76433-610
76433-612
76433-614
76433-616
76433-618
76433-620
76433-622
76433-624
76433-626
76433-628
76433-630
76433-632
76433-634
76433-636
76433-638
76433-640
76433-642
76433-644
76433-646
76433-648
76433-650
76433-652
76433-654
76433-656
76433-658
76433-660
76433-662
76433-664
76433-666
76433-668
76433-670
76433-672
76433-674
76433-676
76433-678
76433-680
76433-682
76433-684
76433-686
76433-688
76433-690
76433-692
76433-694
76433-696
76433-698
76433-700
76433-702
76433-704
76433-706
76433-708
76433-710
76433-712
76433-714
76433-716
76433-718
76433-720
76433-722
76433-724
76433-726
76433-728
76433-730
76433-732
76433-734
76433-736
76433-738
76433-740
76433-742
76433-744
76433-746
76433-748
76433-750
76433-752
76433-754
76433-756
76433-758
76433-760
76433-762
76433-764
76433-766
76433-768
76433-770
76433-772
76433-774
76433-776
76433-778
76433-780
76433-782
76433-784
76433-786
76433-788
76433-790
76433-792
76433-794
76433-796
76433-798
76433-800
76433-802
76433-804
76433-806
76433-808
76433-810
76433-812
76433-814
76433-816
76433-818
76433-820
76433-822
76433-824
76433-826
76433-828
76433-830
76433-832
76433-834
76433-836
76433-838
76433-840
76433-842
76433-844
76433-846
76433-848
76433-850
76434-120
76434-122
76434-124
76434-126
76434-128
76434-130
76434-132
76434-134
76434-136
76434-138
76434-140
76434-142
76434-144
76434-146
76434-148
76434-150
76434-152
Avantor® Hichrom HI-WAX MS, GC Columns
Chromatography Columns
SPECIFICATIONS
Phase name | HI-WAX MS |
Environmentally Preferable | |
Phase composition | Polyethylene Glycol (PEG) - low bleeding |
Polarity | Polar stationary phase |
Crossbond | Yes |
Temperature limit | 270 °C |
USP code | G14, G15, G16 |