Avantor® Hichrom HI-WAX MS, GC Columns
Supplier: VWR International
76431-440 76431-442 76431-444 76431-446 76433-594 76433-596 76433-598 76433-600 76433-602 76433-604 76433-606 76433-608 76433-610 76433-612 76433-614 76433-616 76433-618 76433-620 76433-622 76433-624 76433-626 76433-628 76433-630 76433-632 76433-634 76433-636 76433-638 76433-640 76433-642 76433-644 76433-646 76433-648 76433-650 76433-652 76433-654 76433-656 76433-658 76433-660 76433-662 76433-664 76433-666 76433-668 76433-670 76433-672 76433-674 76433-676 76433-678 76433-680 76433-682 76433-684 76433-686 76433-688 76433-690 76433-692 76433-694 76433-696 76433-698 76433-700 76433-702 76433-704 76433-706 76433-708 76433-710 76433-712 76433-714 76433-716 76433-718 76433-720 76433-722 76433-724 76433-726 76433-728 76433-730 76433-732 76433-734 76433-736 76433-738 76433-740 76433-742 76433-744 76433-746 76433-748 76433-750 76433-752 76433-754 76433-756 76433-758 76433-760 76433-762 76433-764 76433-766 76433-768 76433-770 76433-772 76433-774 76433-776 76433-778 76433-780 76433-782 76433-784 76433-786 76433-788 76433-790 76433-792 76433-794 76433-796 76433-798 76433-800 76433-802 76433-804 76433-806 76433-808 76433-810 76433-812 76433-814 76433-816 76433-818 76433-820 76433-822 76433-824 76433-826 76433-828 76433-830 76433-832 76433-834 76433-836 76433-838 76433-840 76433-842 76433-844 76433-846 76433-848 76433-850 76434-120 76434-122 76434-124 76434-126 76434-128 76434-130 76434-132 76434-134 76434-136 76434-138 76434-140 76434-142 76434-144 76434-146 76434-148 76434-150 76434-152
76431-440EA 1706.51 CAD
76431-440 76431-442 76431-444 76431-446 76433-594 76433-596 76433-598 76433-600 76433-602 76433-604 76433-606 76433-608 76433-610 76433-612 76433-614 76433-616 76433-618 76433-620 76433-622 76433-624 76433-626 76433-628 76433-630 76433-632 76433-634 76433-636 76433-638 76433-640 76433-642 76433-644 76433-646 76433-648 76433-650 76433-652 76433-654 76433-656 76433-658 76433-660 76433-662 76433-664 76433-666 76433-668 76433-670 76433-672 76433-674 76433-676 76433-678 76433-680 76433-682 76433-684 76433-686 76433-688 76433-690 76433-692 76433-694 76433-696 76433-698 76433-700 76433-702 76433-704 76433-706 76433-708 76433-710 76433-712 76433-714 76433-716 76433-718 76433-720 76433-722 76433-724 76433-726 76433-728 76433-730 76433-732 76433-734 76433-736 76433-738 76433-740 76433-742 76433-744 76433-746 76433-748 76433-750 76433-752 76433-754 76433-756 76433-758 76433-760 76433-762 76433-764 76433-766 76433-768 76433-770 76433-772 76433-774 76433-776 76433-778 76433-780 76433-782 76433-784 76433-786 76433-788 76433-790 76433-792 76433-794 76433-796 76433-798 76433-800 76433-802 76433-804 76433-806 76433-808 76433-810 76433-812 76433-814 76433-816 76433-818 76433-820 76433-822 76433-824 76433-826 76433-828 76433-830 76433-832 76433-834 76433-836 76433-838 76433-840 76433-842 76433-844 76433-846 76433-848 76433-850 76434-120 76434-122 76434-124 76434-126 76434-128 76434-130 76434-132 76434-134 76434-136 76434-138 76434-140 76434-142 76434-144 76434-146 76434-148 76434-150 76434-152
Avantor® Hichrom HI-WAX MS, GC Columns
Chromatography Columns
Very popular PEG high polarity MS phase for a wide range of applications, including (for example): essential oils, flavors and fragrances, alkenols. HI-WAX MS is an MS grade low-bleeding crossbonded column.
  • Phase composition: Crossbonded Polyethylene Glycol (PEG) - MS grade
  • Long lifetime
Maximum temperature limits may change depending on the film thickness.