Avantor® Hichrom HI-5 MS Xil, GC Columns
Supplier: VWR International
76431-236 76431-238 76431-240 76431-242 76431-244 76431-246 76431-248 76431-250 76431-252 76431-254 76431-256 76431-258 76433-114 76433-116 76433-118 76433-120 76433-122 76433-124 76433-126 76433-128 76433-130 76433-132 76433-134 76433-136 76433-138 76433-140 76433-142 76433-144 76433-146 76433-148 76433-150 76433-152 76433-154 76433-156 76433-158 76433-160 76433-162 76433-164 76433-166 76433-168 76433-170 76433-172 76433-174 76433-176 76433-178 76433-180 76433-182 76433-184 76433-186 76433-188 76433-190 76433-192 76433-194 76433-196 76433-198 76433-200 76433-202 76433-204 76433-206 76433-208 76433-210 76433-212 76433-214 76433-216 76433-218 76433-220 76433-222 76433-224 76433-226 76433-228 76433-230 76433-232 76433-234 76433-236 76433-238 76433-240 76433-242 76433-244 76433-246 76433-248 76433-250 76433-252 76433-254 76433-256 76433-258 76433-260 76433-262 76433-264 76433-266 76433-268 76433-270 76433-272 76433-274 76433-276 76433-278 76433-280 76433-282 76433-284 76433-286 76433-288 76433-290 76433-292 76433-294 76433-296 76433-298 76433-300 76433-302 76433-304 76433-306 76433-308 76433-310 76433-312 76433-314 76433-316 76433-318 76433-320 76433-322 76433-324 76433-326 76433-328 76433-330 76433-332 76433-334 76433-336 76433-338 76433-340 76433-342 76433-344 76433-346 76433-348 76433-350 76434-044 76434-046 76434-048 76434-050 76434-052 76434-054 76434-056 76434-058 76434-060 76434-062 76434-064 76434-066 76434-068 76434-070 76434-072 76434-074 76434-076
76431-236EA 2258.4 CAD
76431-236 76431-238 76431-240 76431-242 76431-244 76431-246 76431-248 76431-250 76431-252 76431-254 76431-256 76431-258 76433-114 76433-116 76433-118 76433-120 76433-122 76433-124 76433-126 76433-128 76433-130 76433-132 76433-134 76433-136 76433-138 76433-140 76433-142 76433-144 76433-146 76433-148 76433-150 76433-152 76433-154 76433-156 76433-158 76433-160 76433-162 76433-164 76433-166 76433-168 76433-170 76433-172 76433-174 76433-176 76433-178 76433-180 76433-182 76433-184 76433-186 76433-188 76433-190 76433-192 76433-194 76433-196 76433-198 76433-200 76433-202 76433-204 76433-206 76433-208 76433-210 76433-212 76433-214 76433-216 76433-218 76433-220 76433-222 76433-224 76433-226 76433-228 76433-230 76433-232 76433-234 76433-236 76433-238 76433-240 76433-242 76433-244 76433-246 76433-248 76433-250 76433-252 76433-254 76433-256 76433-258 76433-260 76433-262 76433-264 76433-266 76433-268 76433-270 76433-272 76433-274 76433-276 76433-278 76433-280 76433-282 76433-284 76433-286 76433-288 76433-290 76433-292 76433-294 76433-296 76433-298 76433-300 76433-302 76433-304 76433-306 76433-308 76433-310 76433-312 76433-314 76433-316 76433-318 76433-320 76433-322 76433-324 76433-326 76433-328 76433-330 76433-332 76433-334 76433-336 76433-338 76433-340 76433-342 76433-344 76433-346 76433-348 76433-350 76434-044 76434-046 76434-048 76434-050 76434-052 76434-054 76434-056 76434-058 76434-060 76434-062 76434-064 76434-066 76434-068 76434-070 76434-072 76434-074 76434-076
Avantor® Hichrom HI-5 MS Xil, GC Columns
Chromatography Columns
Silphenylene based stationary phase (selectivity equivalent to 5MS columns) to ensure minimum bleeding level at high temperatures. General purpose low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 MS Xil is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
  • Phase composition: Crossbonded Silphenylene - Methyl Polysiloxane (5MS column selectivity) - MS grade
  • Long lifetime
Max temperature limits mayMore Product Information