Avantor® Hichrom HI-5 MS, GC Columns
Supplier: VWR International
76431-188 76431-190 76431-192 76431-194 76431-196 76431-198 76431-200 76431-202 76431-204 76431-206 76431-208 76431-210 76432-638 76432-640 76432-642 76432-644 76432-646 76432-648 76432-650 76432-652 76432-654 76432-656 76432-658 76432-660 76432-662 76432-664 76432-666 76432-668 76432-670 76432-672 76432-674 76432-676 76432-678 76432-680 76432-682 76432-684 76432-686 76432-688 76432-690 76432-692 76432-694 76432-696 76432-698 76432-700 76432-702 76432-704 76432-706 76432-708 76432-710 76432-712 76432-714 76432-716 76432-718 76432-720 76432-722 76432-724 76432-726 76432-728 76432-730 76432-732 76432-734 76432-736 76432-738 76432-740 76432-742 76432-744 76432-746 76432-748 76432-750 76432-752 76432-754 76432-756 76432-758 76432-760 76432-762 76432-764 76432-766 76432-768 76432-770 76432-772 76432-774 76432-776 76432-778 76432-780 76432-782 76432-784 76432-786 76432-788 76432-790 76432-792 76432-794 76432-796 76432-798 76432-800 76432-802 76432-804 76432-806 76432-808 76432-810 76432-812 76432-814 76432-816 76432-818 76432-820 76432-822 76432-824 76432-826 76432-828 76432-830 76432-832 76432-834 76432-836 76432-838 76432-840 76432-842 76432-844 76432-846 76432-848 76432-850 76432-852 76432-854 76432-856 76432-858 76432-860 76432-862 76432-864 76432-866 76432-868 76432-870 76432-872 76432-874 76433-976 76433-978 76433-980 76433-982 76433-984 76433-986 76433-988 76433-990 76433-992 76433-994 76433-996 76433-998 76434-000 76434-002 76434-004 76434-006 76434-008
76431-188EA 2258.4 CAD
76431-188 76431-190 76431-192 76431-194 76431-196 76431-198 76431-200 76431-202 76431-204 76431-206 76431-208 76431-210 76432-638 76432-640 76432-642 76432-644 76432-646 76432-648 76432-650 76432-652 76432-654 76432-656 76432-658 76432-660 76432-662 76432-664 76432-666 76432-668 76432-670 76432-672 76432-674 76432-676 76432-678 76432-680 76432-682 76432-684 76432-686 76432-688 76432-690 76432-692 76432-694 76432-696 76432-698 76432-700 76432-702 76432-704 76432-706 76432-708 76432-710 76432-712 76432-714 76432-716 76432-718 76432-720 76432-722 76432-724 76432-726 76432-728 76432-730 76432-732 76432-734 76432-736 76432-738 76432-740 76432-742 76432-744 76432-746 76432-748 76432-750 76432-752 76432-754 76432-756 76432-758 76432-760 76432-762 76432-764 76432-766 76432-768 76432-770 76432-772 76432-774 76432-776 76432-778 76432-780 76432-782 76432-784 76432-786 76432-788 76432-790 76432-792 76432-794 76432-796 76432-798 76432-800 76432-802 76432-804 76432-806 76432-808 76432-810 76432-812 76432-814 76432-816 76432-818 76432-820 76432-822 76432-824 76432-826 76432-828 76432-830 76432-832 76432-834 76432-836 76432-838 76432-840 76432-842 76432-844 76432-846 76432-848 76432-850 76432-852 76432-854 76432-856 76432-858 76432-860 76432-862 76432-864 76432-866 76432-868 76432-870 76432-872 76432-874 76433-976 76433-978 76433-980 76433-982 76433-984 76433-986 76433-988 76433-990 76433-992 76433-994 76433-996 76433-998 76434-000 76434-002 76434-004 76434-006 76434-008
Avantor® Hichrom HI-5 MS, GC Columns
Chromatography Columns
Really versatile general purpose low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 MS is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
  • Phase composition: crossbonded 5% phenyl - 95% methyl polysiloxane - MS grade
  • Long lifetime
Max temperature limits may change depending on the film thickness.