Anti-CD7 Mouse Monoclonal Antibody [clone: 4H9/CD7] (PE (Phycoerythrin))

Supplier: Biolegend
395603 395604
76422-188EA 167.48 CAD
76422-188 76422-190
Anti-CD7 Mouse Monoclonal Antibody [clone: 4H9/CD7] (PE (Phycoerythrin))
Antibodies
Order Now

SPECIFICATIONS

Antigen symbol CD7
Environmentally Preferable
Antigen synonyms gp40
Clonality Monoclonal
Conjugation PE (Phycoerythrin)
Clone 4H9/CD7
Reactivity Human
Host Mouse
Isotype IgG2a kappa
Flow cytometry Yes
Immunogen T-ALL blast cells
Purification The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions.
Storage buffer Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
Storage temperature 2...8 °C
Tested applications FC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR