PolarSafe® Cooling Transport Bags, Cole Parmer
Supplier: Antyila Scientific
03726-00 03726-01 03726-02 03726-03 03726-32 03726-33 03726-34 03726-35 03726-36 03726-37 03726-38 03726-39
76420-926EA 368.23 CAD
76420-926 76420-928 76420-930 76420-932 76420-940 76420-942 76420-944 76420-946 76420-948 76420-950 76420-952 76420-954
PolarSafe® Cooling Transport Bags, Cole Parmer
Boxes Cooling/Freezing Boxes and Bags
These insulated nylon bags provide an easy solution for transporting your samples.
  • Maintain specific temperature ranges for extended periods of time for improved sample protection
  • Get the temperature protection you need with choice of durable transport bags or boxes
  • Modular, color-coded cooling packs allow flexible configurations for different-sized samples
  • Ensure sample integrity with optional security sealing ties
Organize cooling packs to suit your sample or choose from one of our prepared kits. Bags have pockets to keep documentation safe duringMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable