Moisture Analyzers, MA100
Supplier: Sartorius
MA100C000115V1 MA100H000115V1
76420-844EA 15539.11 CAD
76420-844 76420-846
Moisture Analyzers, MA100
Analysers Moisture Analysers
The MA100 is an accurate and configurable model with a user friendly keyboard and software prompts to easily access all the modes of operation and data input.
  • SPRM- enables the operating parameters to be adapted to the results of an available reference method and be saved as a drying routine
  • Access to sample chamber by motorized cover incorporating the heating unit that reduces interfering effects
  • Password protection of parameters
  • Built-in calibration weight
  • Fully automatic endpoint determination, continuously monitoring the drying curve and stopping the
More Product Information