FlaskScrubber® Glassware Washers, Labconco®
Supplier: Labconco
411001000 411001010 411001013 411101000 411101010 411101013 412001000 412001010 412001013 412101000 412101010 412101013
76420-708EA 24671.7 CAD
76420-708 76420-710 76420-712 76420-714 76420-716 76420-718 76420-720 76420-722 76420-724 76420-726 76420-728 76420-730
FlaskScrubber® Glassware Washers, Labconco®
Glassware Washers
Versatile and efficient glassware washers designed for efficient and consistent washing of narrow-neck glassware and general labware.
  • Touch screen display with intuitive CleanWorks OS software
  • Flexible design allows for use of spindle injection and standard racks
  • Maximum wash temperature of 93 °C removes tough contaminants
  • Advanced wash pump maximizes water circulation up to 112 gal/min (424 liters/min)
  • Standard forced air spindle drying with SpeedVent technology
  • Durable Type 304 stainless steel chamber and racks
These glassware washers are designedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Waste Reducing,Water Efficient