Avantor® ACE® Excel® Glycan, Analytical HPLC/UHPLC Columns
Supplier: Avantor
EXL-1716-1002 EXL-1116-1002 EXL-1116-1502 EXL-1116-1546
76419-556EA 3727.06 CAD
76419-556 76419-558 76419-560 76419-562 76419-564 76419-566 76419-568 76419-570 76419-572 76419-574
Avantor® ACE® Excel® Glycan, Analytical HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® Glycan is a high performance stationary phase designed for the analysis of 2-aminobenzamide labelled glycans by hydrophilic interaction liquid chromatography (HILIC). These ultra-inert, high efficiency columns provide excellent column lifetime and reproducibility with buffers and mobile phases typically utilized for glycan analysis. Two particle sizes are available to enable both high resolution and rapid separations on HPLC and UHPLC instrumentation.
  • High efficiency 1.7 and 3 µm particle sizes for high resolution glycan separations
  • Base
More Product Information