Protector® Airo™ Filtered Fume Hoods, Labconco Corporation
Supplier: Labconco
184300200 184300201 184300202 184300203 184300220 184300221 184300400 184300401 184300402 184300403 184300420 184300421 184300600 184300601 184300602 184300603 184300620 184300621 184300800 184300801 184300802 184300803 184300820 184300821 184310200 184310201 184310202 184310203 184310220 184310221 184310400 184310401 184310402 184310403 184310420 184310421 184310600 184310601 184310602 184310603 184310620 184310621 184310800 184310801 184310802 184310803 184310820 184310821 184410200 184410201 184410202 184410203 184410220 184410221 184410400 184410401 184410402 184410403 184410420 184410421 184410600 184410601 184410602 184410603 184410620 184410621 184410800 184410801 184410802 184410803 184410820 184410821
76419-366EA 31718.5 CAD
76419-366 76419-368 76419-370 76419-372 76419-374 76419-376 76419-378 76419-380 76419-382 76419-384 76419-386 76419-388 76419-390 76419-392 76419-394 76419-396 76419-398 76419-400 76419-402 76419-404 76419-406 76419-408 76419-410 76419-412 76419-414 76419-416 76419-418 76419-420 76419-422 76419-424 76419-426 76419-428 76419-430 76419-432 76419-434 76419-436 76419-438 76419-440 76419-442 76419-444 76419-446 76419-448 76419-450 76419-452 76419-454 76419-456 76419-458 76419-460 76419-462 76419-464 76419-466 76419-468 76419-470 76419-472 76419-474 76419-476 76419-478 76419-480 76419-482 76419-484 76419-486 76419-488 76419-490 76419-492 76419-494 76419-496 76419-498 76419-500 76419-502 76419-504 76419-506 76419-508
Protector® Airo™ Filtered Fume Hoods, Labconco Corporation
Fume Hoods and Enclosures Ductless Fume Hoods
Fully-featured, energy-efficient fume hoods provide an environmentally-friendly solution to ducted fume hoods while simplifying installation due to their compact footprint.
  • Fully featured fume hoods
  • Ducting is not required
  • Energy efficient
  • Environmentally friendly
  • Uses Erlab's Neutrodine Unisorb filtration
  • Four filter combinations available
The Protector® Airo™ Filtered Fume Hood has a compact design that avoids potential challenges of existing laboratory conditions while still providing space, utility and options of a traditional hood. It featuresMore Product Information