Protector® Echo™ Floor-Mounted Filtered Fume Hoods, 55.7" Depth, Labconco Corporation
Supplier: Labconco
183437200 183437201 183437202 183437220 183437221 183437400 183437401 183437402 183437420 183437421 183437600 183437601 183437602 183437620 183437621 183437800 183437801 183437802 183437820 183437821 183537200 183537201 183537202 183537220 183537221 183537400 183537401 183537402 183537420 183537421 183537600 183537601 183537602 183537620 183537621 183537800 183537801 183537802 183537820 183537821 183637200 183637201 183637202 183637220 183637221 183637400 183637401 183637402 183637420 183637421 183637600 183637601 183637602 183637620 183637621 183637800 183637801 183637802 183637820 183637821 183837200 183837201 183837202 183837220 183837221 183837400 183837401 183837402 183837420 183837421 183837600 183837601 183837602 183837620 183837621 183837800 183837801 183837802 183837820 183837821
76418-966EA 60466.6 CAD
76418-966 76418-968 76418-970 76418-972 76418-974 76418-976 76418-978 76418-980 76418-982 76418-984 76418-986 76418-988 76418-990 76418-992 76418-994 76418-996 76418-998 76419-000 76419-002 76419-004 76419-086 76419-088 76419-090 76419-092 76419-094 76419-096 76419-098 76419-100 76419-102 76419-104 76419-106 76419-108 76419-110 76419-112 76419-114 76419-116 76419-118 76419-120 76419-122 76419-124 76419-206 76419-208 76419-210 76419-212 76419-214 76419-216 76419-218 76419-220 76419-222 76419-224 76419-226 76419-228 76419-230 76419-232 76419-234 76419-236 76419-238 76419-240 76419-242 76419-244 76419-326 76419-328 76419-330 76419-332 76419-334 76419-336 76419-338 76419-340 76419-342 76419-344 76419-346 76419-348 76419-350 76419-352 76419-354 76419-356 76419-358 76419-360 76419-362 76419-364
Protector® Echo™ Floor-Mounted Filtered Fume Hoods, 55.7" Depth, Labconco Corporation
Fume Hoods and Enclosures Ductless Fume Hoods
Fully-featured, energy-efficient fume hoods provide an environmentally-friendly solution to ducted fume hoods while simplifying installation. Ideal for oversized equipment.
  • Fully featured fume hood
  • Ducting is not required
  • Energy efficient
  • Environmentally friendly
  • Uses Erlab's Neutrodine Unisorb filtration
  • Four filter combinations available
  • 37.7" and 43.7" (958 and 1110 mm) depths are also available
The Protector® Echo™ Floor-Mounted Filtered Fume Hood combines the trusted filtration of Erlab’s Neutrodine® Unisorb Filter with the quality, innovationMore Product Information