Protector® Echo™ Floor-Mounted Filtered Fume Hoods, 43.7" Depth, Labconco Corporation
Supplier: Labconco
183427200 183427201 183427202 183427220 183427221 183427400 183427401 183427402 183427420 183427421 183427600 183427601 183427602 183427620 183427621 183427800 183427801 183427802 183427820 183427821 183527200 183527201 183527202 183527220 183527221 183527400 183527401 183527402 183527420 183527421 183527600 183527601 183527602 183527620 183527621 183527800 183527801 183527802 183527820 183527821 183627200 183627201 183627202 183627220 183627221 183627400 183627401 183627402 183627420 183627421 183627600 183627601 183627602 183627620 183627621 183627800 183627801 183627802 183627820 183627821 183827200 183827201 183827202 183827220 183827221 183827400 183827401 183827402 183827420 183827421 183827600 183827601 183827602 183827620 183827621 183827800 183827801 183827802 183827820 183827821
76418-926EA 59968.9 CAD
76418-926 76418-928 76418-930 76418-932 76418-934 76418-936 76418-938 76418-940 76418-942 76418-944 76418-946 76418-948 76418-950 76418-952 76418-954 76418-956 76418-958 76418-960 76418-962 76418-964 76419-046 76419-048 76419-050 76419-052 76419-054 76419-056 76419-058 76419-060 76419-062 76419-064 76419-066 76419-068 76419-070 76419-072 76419-074 76419-076 76419-078 76419-080 76419-082 76419-084 76419-166 76419-168 76419-170 76419-172 76419-174 76419-176 76419-178 76419-180 76419-182 76419-184 76419-186 76419-188 76419-190 76419-192 76419-194 76419-196 76419-198 76419-200 76419-202 76419-204 76419-286 76419-288 76419-290 76419-292 76419-294 76419-296 76419-298 76419-300 76419-302 76419-304 76419-306 76419-308 76419-310 76419-312 76419-314 76419-316 76419-318 76419-320 76419-322 76419-324
Protector® Echo™ Floor-Mounted Filtered Fume Hoods, 43.7" Depth, Labconco Corporation
Fume Hoods and Enclosures Ductless Fume Hoods
Fully-featured, energy-efficient fume hoods provide an environmentally-friendly solution to ducted fume hoods while simplifying installation. Ideal for oversized equipment.
  • Fully featured fume hood
  • Ducting is not required
  • Energy efficient
  • Environmentally friendly
  • Uses Erlab's Neutrodine Unisorb filtration
  • Four filter combinations available
  • 37.7" and 55.7" depths also available
The Protector® Echo™ Floor-Mounted Filtered Fume Hood combines the trusted filtration of Erlab’s Neutrodine® Unisorb Filter with the quality, innovation and energy-efficientMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Sustainable Materials,Energy Efficient