Protector® Echo™ Floor-Mounted Filtered Fume Hoods, 37.7" Depth, Labconco Corporation
Supplier: Labconco
183417200 183417201 183417202 183417220 183417221 183417400 183417401 183417402 183417420 183417421 183417600 183417601 183417602 183417620 183417621 183417800 183417801 183417802 183417820 183417821 183517200 183517201 183517202 183517220 183517221 183517400 183517401 183517402 183517420 183517421 183517600 183517601 183517602 183517620 183517621 183517800 183517801 183517802 183517820 183517821 183617200 183617201 183617202 183617220 183617221 183617400 183617401 183617402 183617420 183617421 183617600 183617601 183617602 183617620 183617621 183617800 183617801 183617802 183617820 183617821 183817200 183817201 183817202 183817220 183817221 183817400 183817401 183817402 183817420 183817421 183817600 183817601 183817602 183817620 183817621 183817800 183817801 183817802 183817820 183817821
76418-886EA 59463.3 CAD
76418-886 76418-888 76418-890 76418-892 76418-894 76418-896 76418-898 76418-900 76418-902 76418-904 76418-906 76418-908 76418-910 76418-912 76418-914 76418-916 76418-918 76418-920 76418-922 76418-924 76419-006 76419-008 76419-010 76419-012 76419-014 76419-016 76419-018 76419-020 76419-022 76419-024 76419-026 76419-028 76419-030 76419-032 76419-034 76419-036 76419-038 76419-040 76419-042 76419-044 76419-126 76419-128 76419-130 76419-132 76419-134 76419-136 76419-138 76419-140 76419-142 76419-144 76419-146 76419-148 76419-150 76419-152 76419-154 76419-156 76419-158 76419-160 76419-162 76419-164 76419-246 76419-248 76419-250 76419-252 76419-254 76419-256 76419-258 76419-260 76419-262 76419-264 76419-266 76419-268 76419-270 76419-272 76419-274 76419-276 76419-278 76419-280 76419-282 76419-284
Protector® Echo™ Floor-Mounted Filtered Fume Hoods, 37.7" Depth, Labconco Corporation
Fume Hoods and Enclosures Ductless Fume Hoods
Fully-featured, energy-efficient fume hoods provide an environmentally-friendly solution to ducted fume hoods while simplifying installation. Ideal for oversized equipment.
  • Fully featured fume hood
  • Ducting is not required
  • Energy efficient
  • Environmentally friendly
  • Uses Erlab's Neutrodine Unisorb filtration
  • Four filter combinations available
  • 43.7" and 55.7" depths also available
The Protector® Echo™ Floor-Mounted Filtered Fume Hood combines the trusted filtration of Erlab’s Neutrodine® Unisorb Filter with the quality, innovation and energy-efficientMore Product Information