Protector® Echo™ Filtered Fume Hoods with Side and Back Windows, Labconco Corporation
Supplier: Labconco
182410200 182410201 182410202 182410220 182410221 182410400 182410401 182410402 182410420 182410421 182410600 182410601 182410602 182410620 182410621 182410800 182410801 182410802 182410820 182410821 182510200 182510201 182510202 182510220 182510221 182510400 182510401 182510402 182510420 182510421 182510600 182510601 182510602 182510620 182510621 182510800 182510801 182510802 182510820 182510821 182610200 182610201 182610202 182610220 182610221 182610400 182610401 182610402 182610420 182610421 182610600 182610601 182610602 182610620 182610621 182610800 182610801 182610802 182610820 182610821 182810200 182810201 182810202 182810220 182810221 182810400 182810401 182810402 182810420 182810421 182810600 182810601 182810602 182810620 182810621 182810800 182810801 182810802 182810820 182810821
76418-726EA 42636.3 CAD
76418-726 76418-728 76418-730 76418-732 76418-734 76418-736 76418-738 76418-740 76418-742 76418-744 76418-746 76418-748 76418-750 76418-752 76418-754 76418-756 76418-758 76418-760 76418-762 76418-764 76418-766 76418-768 76418-770 76418-772 76418-774 76418-776 76418-778 76418-780 76418-782 76418-784 76418-786 76418-788 76418-790 76418-792 76418-794 76418-796 76418-798 76418-800 76418-802 76418-804 76418-806 76418-808 76418-810 76418-812 76418-814 76418-816 76418-818 76418-820 76418-822 76418-824 76418-826 76418-828 76418-830 76418-832 76418-834 76418-836 76418-838 76418-840 76418-842 76418-844 76418-846 76418-848 76418-850 76418-852 76418-854 76418-856 76418-858 76418-860 76418-862 76418-864 76418-866 76418-868 76418-870 76418-872 76418-874 76418-876 76418-878 76418-880 76418-882 76418-884
Protector® Echo™ Filtered Fume Hoods with Side and Back Windows, Labconco Corporation
Fume Hoods and Enclosures Ductless Fume Hoods
Fully-featured, energy-efficient fume hoods provide an environmentally-friendly solution to ducted fume hoods while simplifying installation.
  • Fully featured fume hood
  • Ducting is not required
  • Energy efficient
  • Environmentally friendly
  • Uses Erlab's Neutrodine Unisorb filtration
  • Four filter combinations available
  • Models with no windows or side windows are also available
The Protector® Echo™ Filtered Fume Hood combines the trusted filtration of Erlab’s Neutrodine® Unisorb Filter with the quality, innovation and energy-efficient fume hood designs ofMore Product Information