Protector® Echo™ Filtered Fume Hoods with Side Windows, Labconco Corporation
Supplier: Labconco
181410200 181410201 181410202 181410220 181410221 181410400 181410401 181410402 181410420 181410421 181410600 181410601 181410602 181410620 181410621 181410800 181410801 181410802 181410820 181410821 181510200 181510201 181510202 181510220 181510221 181510400 181510401 181510402 181510420 181510421 181510600 181510601 181510602 181510620 181510621 181510800 181510801 181510802 181510820 181510821 181610200 181610201 181610202 181610220 181610221 181610400 181610401 181610402 181610420 181610421 181610600 181610601 181610602 181610620 181610621 181610800 181610801 181610802 181610820 181610821 181810200 181810201 181810202 181810220 181810221 181810400 181810401 181810402 181810420 181810421 181810600 181810601 181810602 181810620 181810621 181810800 181810801 181810802 181810820 181810821
76418-566EA 41625.1 CAD
76418-566 76418-568 76418-570 76418-572 76418-574 76418-576 76418-578 76418-580 76418-582 76418-584 76418-586 76418-588 76418-590 76418-592 76418-594 76418-596 76418-598 76418-600 76418-602 76418-604 76418-606 76418-608 76418-610 76418-612 76418-614 76418-616 76418-618 76418-620 76418-622 76418-624 76418-626 76418-628 76418-630 76418-632 76418-634 76418-636 76418-638 76418-640 76418-642 76418-644 76418-646 76418-648 76418-650 76418-652 76418-654 76418-656 76418-658 76418-660 76418-662 76418-664 76418-666 76418-668 76418-670 76418-672 76418-674 76418-676 76418-678 76418-680 76418-682 76418-684 76418-686 76418-688 76418-690 76418-692 76418-694 76418-696 76418-698 76418-700 76418-702 76418-704 76418-706 76418-708 76418-710 76418-712 76418-714 76418-716 76418-718 76418-720 76418-722 76418-724
Protector® Echo™ Filtered Fume Hoods with Side Windows, Labconco Corporation
Fume Hoods and Enclosures Ductless Fume Hoods
Fully-featured, energy-efficient fume hood provides an environmentally-friendly solution to ducted fume hoods while simplifying installation.
  • Fully featured fume hood
  • Ducting is not required
  • Energy efficient
  • Environmentally friendly
  • Uses Erlab's Neutrodine Unisorb filtration
  • Four filter combinations available
  • Models with no windows or back windows also available
The Protector® Echo™ Filtered Fume Hood combines the trusted filtration of Erlab’s Neutrodine® Unisorb Filter with the quality, innovation and energy-efficient fume hood designs of Labconco'sMore Product Information