Protector® Echo™ Filtered Fume Hoods with No Windows, Labconco Corporation
Supplier: Labconco
180410200 180410201 180410202 180410220 180410221 180410400 180410401 180410402 180410420 180410421 180410600 180410601 180410602 180410620 180410621 180410800 180410801 180410802 180410820 180410821 180510200 180510201 180510202 180510220 180510221 180510400 180510401 180510402 180510420 180510421 180510600 180510601 180510602 180510620 180510621 180510800 180510801 180510802 180510820 180510821 180610200 180610201 180610202 180610220 180610221 180610400 180610401 180610402 180610420 180610421 180610600 180610601 180610602 180610620 180610621 180610800 180610801 180610802 180610820 180610821 180810200 180810201 180810202 180810220 180810221 180810400 180810401 180810402 180810420 180810421 180810600 180810601 180810602 180810620 180810621 180810800 180810801 180810802 180810820 180810821
76418-406EA 39658 CAD
76418-406 76418-408 76418-410 76418-412 76418-414 76418-416 76418-418 76418-420 76418-422 76418-424 76418-426 76418-428 76418-430 76418-432 76418-434 76418-436 76418-438 76418-440 76418-442 76418-444 76418-446 76418-448 76418-450 76418-452 76418-454 76418-456 76418-458 76418-460 76418-462 76418-464 76418-466 76418-468 76418-470 76418-472 76418-474 76418-476 76418-478 76418-480 76418-482 76418-484 76418-486 76418-488 76418-490 76418-492 76418-494 76418-496 76418-498 76418-500 76418-502 76418-504 76418-506 76418-508 76418-510 76418-512 76418-514 76418-516 76418-518 76418-520 76418-522 76418-524 76418-526 76418-528 76418-530 76418-532 76418-534 76418-536 76418-538 76418-540 76418-542 76418-544 76418-546 76418-548 76418-550 76418-552 76418-554 76418-556 76418-558 76418-560 76418-562 76418-564
Protector® Echo™ Filtered Fume Hoods with No Windows, Labconco Corporation
Fume Hoods and Enclosures Ductless Fume Hoods
Fully-featured, energy-efficient fume hoods provide an environmentally-friendly solution to ducted fume hoods while simplifying installation.
  • Fully featured fume hoods
  • Ducting is not required
  • Energy efficient
  • Environmentally friendly
  • Uses Erlab's Neutrodine Unisorb filtration
  • Four filter combinations available
  • Models with windows are also available
The Protector® Echo™ Filtered Fume Hood combines the trusted filtration of Erlab’s Neutrodine® Unisorb Filter with the quality, innovation and energy-efficient fume hood designs of Labconco's unparalleledMore Product Information