Caliber™ Type VII Gauze Sponges, DUKAL™ Corporation
Supplier: Dukal
C5119 C5129 C5139 C517 C5189 C82082 C82122 C83082 C84088 C84166 C84688
76407-978BX 10.84 CAD
76407-978 76407-980 76407-982 76407-984 76407-986 76407-988 76407-990 76407-992 76407-994 76407-996 76407-998 76408-000
Caliber™ Type VII Gauze Sponges, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Gauze sponges are designed for use in a variety of applications, including general wound care, dressings, prepping, wound packing, and debriding.
  • 100% woven cotton
  • No natural rubber latex
  • Available in sterile and non sterile formats
Highly absorbent sponges offer exceptional clinical performance.
Certifications: Meets USP Type VII requirements.