Cohesive Bandages, DUKAL™ Corporation
Supplier: Dukal
8015AS 8025AS 8046PLLF 8155AS 8155DB 8155T 8155W 8156ASLF 8156TLF
76407-944BX 65.36 CAD
76407-944 76407-946 76407-948 76407-950 76407-952 76407-954 76407-956 76407-958 76407-960 76407-962
Cohesive Bandages, DUKAL™ Corporation
First Aid Supplies Wound Dressings
A self-adherent elastic bandage that provides consistent support.
  • Maintains even compression and will not slip in use
  • Self-adherent design eliminates the need for pins or clips
  • Easily torn by hand – no scissors required
  • Conformable bandages will fit to difficult body contours and movement
  • Lightweight and porous for comfort and breathability
  • Easy to apply and remove – an excellent alternative to tape
  • Available in latex and latex-free formats
Common applications include use as a support wrap for sprains and strains, compression bandage, overwrap forMore Product Information