LiChrospher® RP-8, HPLC Columns
Supplier: Avantor
LISPRP8-5-10C5 LISPRP8-5-125A LISPRP8-5-250A LISPRP8-5125AF LISPRP8-5250AF LISPRP8510C5SK
76400-648EA 1431.26 CAD
76400-648 76400-650 76400-652 76400-654 76400-656 76400-658 76400-660 76400-662 76400-664 76400-666 76400-668 76400-676 76400-678 76400-680 76400-682 76400-684 76400-686 76400-688
LiChrospher® RP-8, HPLC Columns
Chromatography Columns
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
  • Spherical porous silica
  • High efficiency packed columns
  • Stainless steel column hardware