Avantor® ACE® UltraCore, HPLC/UHPLC Columns, Method Development Kits, 2.5 µm
Supplier: Avantor
MDKU-25-0502U MDKU-25-0503U MDKU-25-0546U MDKU-25-1002U MDKU-25-1003U MDKU-25-1046U MDKU-25-1502U MDKU-25-1503U MDKU-25-1546U
76397-390EA 2451.84 CAD
76397-390 76397-392 76397-394 76397-404 76397-406 76397-408 76397-432 76397-434 76397-436
Avantor® ACE® UltraCore, HPLC/UHPLC Columns, Method Development Kits, 2.5 µm
Chromatography Columns
Avantor® ACE® Method Development Kits (MDK) are designed to maximize selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness. A complete range of kits for all requirements is available, including porous, solid core, HILIC and bioanalytical 300 Å column kits. These kits group together the essential columnMore Product Information