Avantor® ACE®, HILIC Method Development Kits, Analytical, HPLC/UHPLC Columns
Supplier: Avantor
MDKH-17-0502U MDKH-17-0503U MDKH-17-1002U MDKH-17-1003U MDKH-3-0502U MDKH-3-0503U MDKH-3-0546U MDKH-3-1002U MDKH-3-1003U MDKH-3-1046U MDKH-3-1502U MDKH-3-1503U MDKH-3-1546U MDKH-3-2502U MDKH-3-2503U MDKH-3-2546U MDKH-5-0502U MDKH-5-0503U MDKH-5-0546U MDKH-5-1002U MDKH-5-1003U MDKH-5-1046U MDKH-5-1502U MDKH-5-1503U MDKH-5-1546U MDKH-5-2502U MDKH-5-2503U MDKH-5-2546U
76397-134EA 5121.97 CAD
76397-134 76397-136 76397-138 76397-140 76397-178 76397-180 76397-182 76397-192 76397-194 76397-196 76397-220 76397-222 76397-224 76397-226 76397-228 76397-230 76397-280 76397-282 76397-284 76397-294 76397-296 76397-298 76397-322 76397-324 76397-326 76397-336 76397-338 76397-340
Avantor® ACE®, HILIC Method Development Kits, Analytical, HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® HILIC method development kits offer all three Avantor® ACE® proprietary HILIC phases (HILIC-A, HILIC-B and HILIC-N) in one kit.
  • Ideal for the separation and retention of polar analytes
  • Compatible with all manufacturers’ HPLC/UHPLC systems
  • A wide range of column lengths and internal diameters are available
  • Stainlesss teel column hardware
These phases offer alternative HILIC selectivity to each other, and have been developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach toMore Product Information