Avantor® ACE®, Extended Method Development Kits, 5 µm, HPLC/UHPLC Columns
Supplier: Avantor
MDKE-5-0502U MDKE-5-0503U MDKE-5-0546U MDKE-5-1002U MDKE-5-1003U MDKE-5-1046U MDKE-5-1502U MDKE-5-1503U MDKE-5-1546U MDKE-5-2502U MDKE-5-2503U MDKE-5-2546U
76397-044EA 2146.48 CAD
76397-044 76397-046 76397-048 76397-058 76397-060 76397-062 76397-086 76397-088 76397-090 76397-100 76397-102 76397-104
Avantor® ACE®, Extended Method Development Kits, 5 µm, HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1.7 and 5 µm ) and complementary phases
  • Analytical IDs in a
More Product Information