Avantor® ACE® HILIC-N, Analytical, HPLC/UHPLC Columns
Supplier: Avantor
HILN-17-0502U HILN-17-0503U HILN-17-1002U HILN-17-1003U HILN-3-0102GD HILN-3-0103GD HILN-3-0303U HILN-3-0502U HILN-3-1002U HILN-3-1003U HILN-3-1502U HILN-3-1503U HILN-3-1546U HILN-3-2503U HILN-3-2546U HILN-3-7503U HILN-5-0103GD HILN-5-1002U HILN-5-1546U HILN-5-2503U HILN-5-2546U
76395-844EA 1537.21 CAD
76395-844 76395-846 76395-848 76395-850 76395-852 76395-854 76395-856 76395-858 76395-860 76395-862 76395-864 76395-866 76395-870 76395-872 76395-874 76395-876 76395-878 76395-888 76395-890 76395-892 76395-902 76395-904 76395-906 76395-916 76395-918 76395-920 76395-930 76395-932 76395-934 76395-944 76395-946 76395-948 76395-950 76395-952 76395-954 76395-956 76395-958 76395-960 76395-970 76395-972 76395-974 76395-976 76395-978 76395-980 76395-982 76395-984 76395-986 76395-988 76395-998 76396-000 76396-002 76396-012 76396-014 76396-016 76396-018 76396-020 76396-022 76396-024 76396-034 76396-036 76396-038 76396-040 76396-042 76396-044 76396-046 76396-056 76396-058 76396-064 76396-066 76396-076 76396-078 76396-080 76396-082 76396-084 76396-086 76396-096 76396-098 76396-100 76396-102 76396-104 76396-106 76396-108 76396-110 76396-112 76396-114 76396-116 76396-118 76396-120 76396-122 76396-132 76396-134 76396-140 76396-142 76396-144
Avantor® ACE® HILIC-N, Analytical, HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® HILIC-N columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are part of a suit of 3 proprietary HILIC columns (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
  • Proprietary polyhydroxy neutral character HILIC phase
  • Compatible with all manufacturers’ HPLC/UHPLC systems
  • Excellent column performance, inertness and robustness
  • Simplified
More Product Information