Avantor® ACE® HILIC-B, Analytical, HPLC/UHPLC Columns
Supplier: Avantor
HILB-3-1046U HILB-3-1502U HILB-3-1503U HILB-3-1546U HILB-3-2546U HILB-5-0502U HILB-17-0203U HILB-17-0302U HILB-17-0303U HILB-17-0502U HILB-17-1002U HILB-17-1003U HILB-17-7502U HILB-3-0302U HILB-3-0303U HILB-3-0502U HILB-3-1002U HILB-3-1003U HILB-5-1502U HILB-5-1546U HILB-5-2546U
76395-616EA 2277.27 CAD
76395-616 76395-626 76395-628 76395-630 76395-640 76395-642 76395-644 76395-646 76395-648 76395-650 76395-652 76395-654 76395-656 76395-666 76395-668 76395-670 76395-680 76395-682 76395-684 76395-694 76395-696 76395-698 76395-708 76395-710 76395-720 76395-540 76395-542 76395-544 76395-546 76395-548 76395-550 76395-552 76395-554 76395-556 76395-558 76395-560 76395-562 76395-570 76395-572 76395-574 76395-584 76395-586 76395-588 76395-598 76395-600 76395-602 76395-612 76395-614 76395-730 76395-732 76395-742 76395-752 76395-754 76395-762 76395-772 76395-774 76395-784 76395-794 76395-796 76395-806 76395-816 76395-818 76395-820 76395-830 76395-832 76395-842
Avantor® ACE® HILIC-B, Analytical, HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® HILIC-B columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are part of a suit of three proprietary HILIC columns (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
  • Proprietary aminopropyl basic character HILIC phase
  • Compatible with all manufacturers’ HPLC/UHPLC systems
  • Excellent column performance, inertness and robustness
More Product Information