Avantor® ACE® Excel® C8, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
ACE-122-0103GD ACE-122-0102GD EXL-122-0202U EXL-122-0246U EXL-122-0502U EXL-122-0503U EXL-122-1046U EXL-122-1246U EXL-122-1546U EXL-122-2502U EXL-122-2546U
76386-636PK 721.06 CAD
76386-636 76386-634 76394-066 76394-068 76394-070 76394-072 76394-074 76394-076 76394-078 76394-080 76394-082 76394-084 76394-086 76394-088 76394-090 76394-092 76394-094 76394-096 76394-098 76394-100 76394-102 76394-104 76394-106 76394-108 76394-110 76394-112
Avantor® ACE® Excel® C8, HPLC/UHPLC Columns, Analytical, 5 µm
Chromatography Columns
These ultra-inert Avantor® ACE® Excel® C8 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Can be used as general purpose starting point column
  • Wide range of particle sizes (2, 3 and 5 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase -
More Product Information