Avantor® ACE® Excel® AQ, HPLC/UHPLC Columns, Analytical, 3 µm
Supplier: Avantor
EXL-116-0202U EXL-116-0302U EXL-116-0303U EXL-116-0502U EXL-116-0503U EXL-116-0546U EXL-116-1002U EXL-116-1003U EXL-116-1046U EXL-116-1203U EXL-116-1502U EXL-116-1503U EXL-116-1546U EXL-116-2502U EXL-116-2503U EXL-116-2546U EXL-116-7503U
76393-584EA 1969.94 CAD
76393-584 76393-586 76393-588 76393-590 76393-592 76393-594 76393-596 76393-598 76393-600 76393-602 76393-604 76393-606 76393-608 76393-610 76393-612 76393-614 76393-616 76393-618 76393-620 76393-622 76393-624 76393-626 76393-628 76393-630 76393-632 76393-634 76393-636
Avantor® ACE® Excel® AQ, HPLC/UHPLC Columns, Analytical, 3 µm
Chromatography Columns
Avantor® ACE® Excel® AQ columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Retains polar compounds in 100% aqueous mobile phase
  • Unique C18 bonded phase with integral polar functionality
  • Ideal for use with fast gradients due to its rapid re-equilibration properties and resistance to retention loss
  • Wide range of particle sizes (2 to 5 µm) and
More Product Information