Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 2 µm
Supplier: Avantor
EXL-1013-0302U EXL-1013-0502U EXL-1013-0503U EXL-1013-1002U EXL-1013-1003U EXL-1013-1046U EXL-1013-1502U EXL-1013-1503U EXL-1013-1546U EXL-1013-7546U ACE-1P13-GD2U ACE-1P13-GD4U
76392-648EA 1361.74 CAD
76392-648 76392-650 76392-652 76392-654 76392-656 76392-658 76392-660 76392-662 76392-664 76392-666 76392-668 76392-670 76392-672 76392-674 76392-676 76392-678 76392-680 76392-682 76392-684 76392-686 76392-688 76392-696 76392-698 76392-700 76408-410 76408-412
Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 2 µm
Chromatography Columns
Avantor® ACE® CN-ES combines CN polar selectivity with enhanced hydrophobicity, enabling the benefits of both interactions to be fully exploited. Extra phase stability and increased column lifetime is provided by an extended alkyl chain spacer between the silica surface and cyano group. Avantor® ACE® CN-ES is suitable for both reversed phase and normal phase separations.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • With hydrophobic plus CN polar selectivity, especially useful for polar molecules
  • Wide range of particle sizes (1
More Product Information