Avantor® ACE® Excel® C18, Analytical UHPLC Columns, 2 µm
Supplier: Avantor
EXL-101-0502U EXL-101-1502U EXL-101-0202U EXL-101-0302U EXL-101-0303U EXL-101-0346U EXL-101-0503U EXL-101-0546U EXL-101-1002U EXL-101-1003U EXL-101-1046U EXL-101-1203U EXL-101-1246U EXL-101-1503U EXL-101-1546U EXL-101-3502U EXL-101-7502U EXL-101-7503U EXL-101-7546U ACE-1P1-GD2U ACE-1P1-GD4U
76382-606EA 2443.06 CAD
76382-606 76382-608 76392-510 76392-512 76392-514 76392-516 76392-518 76392-520 76392-528 76392-530 76392-538 76392-540 76392-554 76392-578 76392-580 76392-582 76392-590 76392-592 76392-690 76392-692 76392-694 76392-702 76392-704 76392-706 76408-394 76408-396
Avantor® ACE® Excel® C18, Analytical UHPLC Columns, 2 µm
Chromatography Columns
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 to 5 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range ofMore Product Information