Avantor® ACE® C18-300, HPLC Columns, 3 µm
Supplier: Avantor
ACE-211-0102GD ACE-211-0103GD ACE-211-0302 ACE-211-05003 ACE-211-0501 ACE-211-0502 ACE-211-0503 ACE-211-0546 ACE2111000075 ACE-211-10003 ACE-211-10005 ACE-211-1002 ACE-211-1003 ACE-211-1004 ACE-211-1046 ACE-211-1204 ACE-211-15003 ACE-211-1502 ACE-211-1503 ACE-211-1504 ACE-211-2502 ACE-211-2546 ACE-211-7503
76389-408PK 494.3 CAD
76389-408 76389-410 76389-412 76389-414 76389-416 76389-418 76389-420 76389-422 76389-424 76389-426 76389-428 76389-430 76389-432 76389-434 76389-436 76389-438 76389-440 76389-442 76389-444 76389-446 76389-448 76389-450 76389-404 76389-406 76389-452 76389-454 76389-456 76389-458 76389-460 76389-462 76389-464 76389-466 76389-468 76389-470 76389-472 76389-474 76389-476 76389-478 76389-480 76389-482 76389-484 76389-486 76389-488 76389-490 76389-492 76389-494 76389-496 76389-498 76389-500 76389-502 76389-504 76389-506 76389-508 76389-510 76389-512 76389-514 76389-516 76389-518 76389-520 76389-522 76389-524 76389-526 76389-528 76389-530 76389-532 76389-534 76389-536 76389-538 76389-540 76389-542 76389-544 76389-546 76389-548 76389-550 76389-552 76389-554
Avantor® ACE® C18-300, HPLC Columns, 3 µm
Chromatography Columns
Avantor® ACE® C18-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
  • Designed for peptide and protein separations
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle
More Product Information