Avantor® ACE® C8, HPLC Columns, 10 µm
Supplier: Avantor
ACE-132-0110GD ACE-132-0510 ACE-132-1046 ACE-132-1546 ACE-132-2504 ACE-132-2546
76388-732PK 429.79 CAD
76388-732 76388-734 76388-736 76388-738 76388-740 76388-742 76388-744 76388-746 76388-748 76388-750 76388-752 76388-754 76388-756 76388-758 76388-760 76388-762 76388-764 76388-770 76388-772 76388-774 76388-776 76388-778 76388-780 76388-782 76388-784 76388-786 76388-788 76388-790 76388-792 76388-794 76388-796 76388-798 76388-800 76388-802 76388-804 76388-806 76388-808 76388-810 76388-812 76388-828 76388-830 76388-832
Avantor® ACE® C8, HPLC Columns, 10 µm
Chromatography Columns
These ultra-inert Avantor® ACE® C8 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 10 µm) and complementary phases
  • Can be used as general purpose starting point column
  • Method validation kits contain three different batches of the same phase - available
More Product Information