Avantor® ACE® C18-AR, HPLC Columns, 3 µm
Supplier: Avantor
ACE-119-0103GD ACE-119-0546 ACE-119-1546 ACE-119-0504 ACE-119-1002 ACE-119-1003 ACE-119-1004 ACE-119-1046 ACE-119-1203 ACE-119-1204 ACE-119-7502 ACE-119-7503 ACE-119-7546 ACE-119-0102GD ACE-119-0202 ACE-119-0203 ACE-119-0302 ACE-119-0303 ACE-119-05003 ACE-119-0501 ACE-119-0502 ACE-119-0503 ACE-119-1502 ACE-119-1503 ACE-119-1504 ACE1191546MVK
76382-566PK 715.8 CAD
76382-566 76382-568 76382-570 76385-550 76385-552 76385-554 76385-556 76385-558 76385-560 76385-562 76385-564 76385-566 76385-568 76385-570 76385-572 76385-574 76385-576 76385-578 76385-580 76385-582 76385-584 76385-586 76385-588 76385-590 76385-592 76385-638 76385-640 76385-642 76385-644 76385-646 76385-648 76385-650 76385-652 76385-654 76385-656 76385-658 76385-506 76385-508 76385-510 76385-512 76385-514 76385-516 76385-518 76385-520 76385-522 76385-524 76385-526 76385-528 76385-530 76385-532 76385-534 76385-536 76385-538 76385-540 76385-542 76385-544 76385-546 76385-548 76385-594 76385-596 76385-598 76385-600 76385-602 76385-604 76385-606 76385-608 76385-610 76385-612 76385-614 76385-616 76385-618 76385-620 76385-626 76385-630 76385-632 76385-634 76385-636
Avantor® ACE® C18-AR, HPLC Columns, 3 µm
Chromatography Columns
The Avantor® ACE® C18-AR phase utilizes a specially developed ligand combining a C18 chain with integral phenyl functionality, thus combining the benefits of both C18 and phenyl characteristics into a single phase. This can be used for ‘standard’ C18 column separations, but is additionally recommended for separations that involve compounds containing aromatic functionality.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 10 µm) and
More Product Information