BRAND® Transferpette® S -8/-12 Multi-Channel Mechanical Pipettes, Variable Volume
Supplier: Brandtech
705900 705912 705910 705908 705906 705920 705932 705930 705928 705926
76383-100EA 1818.18 CAD
76383-100 76383-102 76383-104 76383-106 76383-108 76383-110 76383-112 76383-114 76383-116 76383-118
BRAND® Transferpette® S -8/-12 Multi-Channel Mechanical Pipettes, Variable Volume
Pipettes Micropipettes
The Transferpette® S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Due to its perfect ergonomics, the Transferpette® S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands.
  • True one-hand operation for right- and left-handed users: Easily set the volume, pipette and eject the tip
  • Color code on volume adjustment wheel, shaft and volume-change protection
  • Low operating forces and a short stroke ensure relaxed pipetting, even during long
More Product Information