Avantor® ACE® C18, HPLC Columns, 3 µm
Supplier: Avantor
ACE-111-0502 ACE-111-0503 ACE-111-0546 ACE-111-1003 ACE-111-1046 ACE-111-1503 ACE-111-1546 ACE-111-7546 ACE-111-0103GD ACE-111-0302 ACE-111-0246 ACE-111-0303 ACE-111-0346 ACE-111-0501 ACE-111-0504 ACE-111-7502 ACE-111-7503 ACE-111-7504 ACE-111-10003 ACE-111-10005 ACE-111-1001 ACE-111-1002 ACE-111-1004 ACE1111046MVK ACE-111-3346 ACE-111-3502 ACE-111-3546 ACE-111-1246 ACE1111500075 ACE-111-15003 ACE-111-15003S ACE-111-1501 ACE-111-1502 ACE-111-1504 ACE1111546MVK ACE111005003GD ACE-111-1202 ACE-111-1203 ACE-111-1204 ACE-111-0101GD ACE-111-0102GD
76382-532EA 2261.42 CAD
76382-532 76382-534 76382-536 76382-538 76382-540 76382-542 76382-544 76382-548 76382-686 76382-688 76384-150 76384-152 76384-154 76384-170 76384-172 76384-174 76384-176 76384-178 76384-180 76384-182 76384-370 76384-196 76384-198 76384-200 76384-202 76384-204 76384-210 76384-212 76384-214 76384-404 76384-406 76384-408 76384-414 76384-424 76384-548 76384-550 76384-552 76384-554 76384-556 76384-558 76384-560 76384-562 76384-564 76384-234 76384-236 76384-238 76384-240 76384-242 76384-244 76384-246 76384-248 76384-250 76384-264 76384-266 76384-470 76384-472 76384-474 76384-476 76384-478 76384-480 76384-482 76384-484 76384-486 76384-320 76384-322 76384-324 76384-326 76384-328 76384-330 76384-332 76384-334 76384-336 76384-338 76384-340 76384-342 76384-344 76384-356 76384-066 76384-302 76384-304 76384-306 76384-308 76384-310 76384-312 76384-314 76384-316 76384-104 76384-106
Avantor® ACE® C18, HPLC Columns, 3 µm
Chromatography Columns
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range ofMore Product Information