Pro-Wipe® 750 Polypropylene/Cellulose Composite Nonwoven Cleanroom Wipes, Berkshire
Supplier: Berkshire
PW750121220 PW750.0404.20 PW750.0909.20
21914-408CS 1181.02 CAD
21914-408 76382-528 76417-126
Pro-Wipe® 750 Polypropylene/Cellulose Composite Nonwoven Cleanroom Wipes, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Pro-Wipe® 750 is a solvent-resistant wipe with exceptional absorbency that makes an excellent upgrade for industrial wipe users. It is composed of a layer of cellulose bonded between two outer layers of polypropylene.
  • Excellent combination of polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Nonwoven wipes with knife-cut edge
  • Smooth, highly absorbent, and durable
  • Low extractable levels and particle counts
  • Chemically compatible with common cleaning and disinfecting solutions
These wipes are engineered for compatibilityMore Product Information