Single-Use High Pressure Transfer Hose with SterilEnz™ Connections
Supplier: AVANTOR FLUID HANDLING, LLC
16SR10P10P60 16SR10P10P120
76379-730EA 965.25 CAD
76379-730 76379-732
Single-Use High Pressure Transfer Hose with SterilEnz™ Connections
Tubing Transfer Tubing
Avantor's high-pressure single-use transfer hose with Avantor SterilEnz™ connections maintain the durability of high-pressure hose products while providing the benefits and flexibility of single-use assemblies.
  • Suitable for elevated temperatures and pressures up to 120 psi
  • 1" inner diameter for high flow rate applications
  • Gamma-irradiated with SAL 10⁻⁶
  • Offered at 5 and 10 foot lengths
  • Gasket affixed to TC face even when disconnected to prevent misalignment
Designed for applications where pressures would exceed normal biopharmaceutical processes.
Order Now
SPECIFICATIONS
Environmentally Preferable