VWR® P2 Balances with Calibration Certificate
Supplier: VWR International
76378-362 76378-364 76378-366 76378-368 76378-370 76378-372 76378-374 76480-262 76480-264 76480-266 76480-268 76480-270
76378-362EA 1903.65 CAD
76378-362 76378-364 76378-366 76378-368 76378-370 76378-372 76378-374 76480-262 76480-264 76480-266 76480-268 76480-270
VWR® P2 Balances with Calibration Certificate
Balances and Scales
VWR® P2-Series/C balances which include an ISO/IEC 17025:2017 Calibration Certificate are the answer for an organizations regulatory and compliance needs.
  • Complete calibration data reported
  • Internal, rechargeable battery
  • Backlit, LCD screen
  • External and internal calibration options
  • RS232 and USB (A and B) communication interfaces
  • Five year warranty
The economical balances include a Certificate of Calibration showing the full testing data from an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory utilizing NIST-traceable weights. TheseMore Product Information